Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Chlorophenylsulphonyl)-3-propylurea ≥99.0% (by titrimetric analysis)
1-(4-Chlorophenylsulphonyl)-3-propylurea ≥99.0% (by titrimetric analysis)
Catalog # TCIAC1220-25G
Supplier:  TCI
CAS Number:  
1-(4-Chlorophenylsulphonyl)-3-propylurea ≥99.0% (by titrimetric analysis)
Catalog # TCIAC1220-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    127.0 to 131.0 deg-C
  • Solubility in Acetone:
    almost transparency

Specification Test Results

About this item

 :
 :276.74 g/mol
Merck Index:14,02186
EINECS:202-314-5
CAS Number:94-20-2
Formula:C₁₀H₁₃ClN₂O₃S
MDL Number:MFCD00079004