

4-Chlorobenzenesulphonic acid hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0606-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Neutralization titration):min. 98.0% (calcd.on anh.substance)
- Solubility in Water:very faint turbidity
- Water:7.5 to 10.0 %
Specification Test Results
About this item
:
:192.61 g/mol
Merck Index:14,02122
EINECS:202-690-0
UN:2585
ADR:8,III
:GHS05
:GHS07
CAS Number:98-66-8
Formula:C₆H₅ClO₃S
MDL Number:MFCD00065342