
S-(2-Benzothiazolyl) (Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)thioacetate ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAB3602-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:350.43 g/mol
EINECS:279-540-6
:GHS07
CAS Number:80756-85-0
Formula:C₁₃H₁₀N₄O₂S₃
MDL Number:MFCD00129148