Order Entry
Czech Republic
ContactUsLinkComponent
NBD-COCl [=4-(N-Chloroformylmethyl-N-methylamino)-7-nitro-2,1,3-benzoxadiazole] ≥92.0% (by titrimetric analysis) for HPLC labeling
Corrosive
NBD-COCl [=4-(N-Chloroformylmethyl-N-methylamino)-7-nitro-2,1,3-benzoxadiazole] ≥92.0% (by titrimetric analysis) for HPLC labeling
Catalog # TCIAA5572-100MG
Supplier:  TCI
CAS Number:  
Corrosive
NBD-COCl [=4-(N-Chloroformylmethyl-N-methylamino)-7-nitro-2,1,3-benzoxadiazole] ≥92.0% (by titrimetric analysis) for HPLC labeling
Catalog # TCIAA5572-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystaline
  • Purity (Argentmetric Titration):
    min. 92.0 %
  • Effect test of HPLC derivatization:
    Effective as labeling agent for Dodecanol

Specification Test Results

About this item

 :
 :270.63 g/mol
Storage Temperature:Freezer
UN:3261
ADR:8,III
 :GHS05
CAS Number:140164-85-8
Formula:C₉H₇ClN₄O₄
MDL Number:MFCD02093432