Order Entry
Czech Republic
ContactUsLinkComponent
2-Anthraceneboronic acid (contains varying amounts of Anhydride)
Irritant
2-Anthraceneboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAA1843-1G
Supplier:  TCI
CAS Number:  
Irritant
2-Anthraceneboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAA1843-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Neutralization titration):
    97.0 to 109.0 %

Specification Test Results

About this item

 :
 :222.05 g/mol
 :GHS07
CAS Number:141981-64-8
Formula:C₁₄H₁₁BO₂
MDL Number:MFCD07784001