

Sodium-4-aminophenylarsonate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0531-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:239.04 g/mol
Merck Index:14,08578
EINECS:204-869-9
UN:2473
ADR:6.1,III
:GHS06
:GHS09
CAS Number:127-85-5
Formula:C₆H₈AsNO₃
MDL Number:MFCD00067431