Acrylamide polymer, M.W. 400000-800000 10% in water (containing small amounts of formalin as fungicide)
Catalog # TCIAA0140-500G
Supplier: TCI
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Molecular weight:400,000 to 800,000
Specification Test Results
About this item
:
Storage Temperature:Refrigerator
CAS Number:9003-05-8
Formula:(CH₂:CHCONH₂)n
MDL Number:MFCD00084392