Order Entry
Czech Republic
ContactUsLinkComponent
Acriflavine ≥70% (by HPLC) (mixture of 3,6-Diamino-10-methylacridinium Chloride and 3,6-Diaminoacridine)
Irritant
Acriflavine ≥70% (by HPLC) (mixture of 3,6-Diamino-10-methylacridinium Chloride and 3,6-Diaminoacridine)
Catalog # TCIAA0134-25G
Supplier:  TCI
CAS Number:  
Irritant
Acriflavine ≥70% (by HPLC) (mixture of 3,6-Diamino-10-methylacridinium Chloride and 3,6-Diaminoacridine)
Catalog # TCIAA0134-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (HPLC):
    min. 70.0 % (as Acriflavine)

Specification Test Results

About this item

 :
 :468.9806 g/mol
Merck Index:14,00123
 :GHS07
CAS Number:8048-52-0
 :65589-70-0
Formula:C₂₇H₂₅ClN₆
MDL Number:MFCD00064307