

1-Cetylpyridinium chloride monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6004-24-6
:
Formula:C₂₁H₃₈ClN·1H₂O
MDL Number:MFCD00149977
:358.01 g/mol
Merck Index:13,02041
Melting Pt:77…83 °C
EINECS:204-593-9
UN:2811
ADR:6.1,I
:M-Clarity™ quality level = MQ200
:GHS06
:GHS09
Storage Temperature:Ambient
Frequently Bought Together