

1-Cetylpyridinium chloride monohydrate, Sigma-Aldrich®
Catalog # SIAL52350-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:358.01 g/mol
Merck Index:13,02041
Melting Pt:77…83 °C
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:204-593-9
UN:2811
:GHS06
:GHS09
Storage Temperature:Ambient
CAS Number:6004-24-6
Formula:C₂₁H₃₈ClN·1H₂O
MDL Number:MFCD00149977