Order Entry
Czech Republic
ContactUsLinkComponent
2-Methoxy-1-methylethyl acetate (PGMEA) stabilised for synthesis, Sigma-Aldrich®
Flammable
2-Methoxy-1-methylethyl acetate (PGMEA) stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.18532.1000
Supplier:  Merck
CAS Number:  
Flammable
2-Methoxy-1-methylethyl acetate (PGMEA) stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.18532.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    209.3 g/l (20 °C)
  • Melting Point:
    -67 °C
  • Molar Mass:
    132.16 g/mol
  • Boiling Point:
    145 - 147 °C (1013 hPa)
  • Vapor Pressure:
    5 hPa (20 °C)
  • Flash Point:
    42 °C
  • Refractive Index:
    1.4078 (20 °C, 589 nm)
  • Explosion Limit:
    1.3 - 10.8 %(V)
  • Density:
    0.97 g/cm3 (20 °C)
  • pH:
    4 (200 g/l, H2O, 20 °C)
  • Ignition Point:
    315 °C
  • Water absorption:
    320 g/kg (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

About this item

 :
 :132.16 g/mol
Boiling Pt:145.8 °C (1013 hPa)
Melting Pt:–67 °C
Density:0.968 g/cm³ (20 °C)
Flash Pt:45 °C
Storage Temperature:Ambient
ADR:3,III
 :M-Clarity™ quality level = MQ200
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:203-603-9
UN:3272
 :GHS02
CAS Number:108-65-6
Formula:C₆H₁₂O₃
MDL Number:MFCD00038500