


Nickel(II) perchlorate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13520-61-1
:
Formula:Cl₂NiO₈·6H₂O
MDL Number:MFCD00149804
:365.68 g/mol
Melting Pt:140 °C
EINECS:237-124-1
UN:1481
ADR:5.1,II
:M-Clarity™ quality level = MQ100
:GHS05
:GHS08
:GHS09
Frequently Bought Together