L(+)-Lysine monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:39665-12-8
:
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MDL Number:MFCD00151035
Melting Pt:225 °C
:M-Clarity™ quality level = MQ200
:164.2 g/mol
Storage Temperature:Refrigerator
EINECS:200-294-2
Frequently Bought Together