

Copper(II) perchlorate hexahydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-46-9
:
Formula:Cl₂CuO₈·6H₂O
MDL Number:MFCD00149666
:370.54
Merck Index:14,2647
Melting Pt:82 °C
Density:2.23
Storage Temperature:Ambient
EINECS:237-391-4
UN:1481
ADR:5.1,II
:GHS03
:GHS07
Related Information
Frequently Bought Together