(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:22323-82-6
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00063239
:132.16 g/mol
Merck Index:12,05232
Boiling Pt:82 °C (1013 hPa)
Density:1.0704 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:244-910-8
Frequently Bought Together