Order Entry
Czech Republic
ContactUsLinkComponent
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
Certificates
undefined
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4330 to 1.4350 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    +10.5° to +11.5° (20°C, 589 nm) (c=5, CH3OH)

Specification Test Results

Related Information

Frequently Bought Together