(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:14347-78-5
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00003213
:132.16 g/mol
Merck Index:12,05232
Boiling Pt:72 °C (1013 hPa)
Density:1.07 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:405-320-8
Frequently Bought Together