Order Entry
Czech Republic
ContactUsLinkComponent
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 96%
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 96%
Catalog # B23037.06
CAS Number:  
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 96%
Catalog # B23037.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :132.16 g/mol
Merck Index:14,5213
Boiling Pt:82…83 °C (14 mmHg)
Density:1.066
Flash Pt:80 °C (176 °F)
Storage Temperature:Refrigerator
EINECS:405-320-8
CAS Number:14347-78-5
Formula:C₆H₁₂O₃
MDL Number:MFCD00003213