Order Entry
Czech Republic
ContactUsLinkComponent
Lanthanum(III) chloride heptahydrate ≥98%, AnalaR NORMAPUR® analytical reagent
Lanthanum(III) chloride heptahydrate ≥98%, AnalaR NORMAPUR® analytical reagent
Catalog # 103433Q
Supplier:  VWR Chemicals
CAS Number:  
undefined
Lanthanum(III) chloride heptahydrate ≥98%, AnalaR NORMAPUR® analytical reagent
Catalog # 103433Q
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Identification:
    Passes test
  • Insolubility in water:
    Max. 0.1 %
  • Ca (Calcium):
    Max. 10 ppm
  • Cu (Copper):
    Max. 1 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Mg (Magnesium):
    Max. 1 ppm
  • Pb (Lead):
    Max. 2 ppm
  • Zn (Zinc):
    Max. 3 ppm

Specification Test Results

About this item

 :
Boiling Pt:< 860 °C (1013 hPa)
Melting Pt:96 °C
 :371.37 g/mol
Storage Temperature:Ambient
EINECS:233-237-5
CAS Number:10025-84-0
Formula:LaCl₃·7H₂O
MDL Number:MFCD00149756