Gallium trinitrate hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:69365-72-6
:
Formula:GaN₃O₉·H₂O
MDL Number:MFCD00149723
:273.75 g/mol
ADR:5.1,III
:M-Clarity™ quality level = MQ200
EINECS:236-815-5
UN:1477
Frequently Bought Together