Order Entry
Czech Republic
ContactUsLinkComponent
Disodium metasilicate pentahydrate, TECHNICAL
Corrosive
Irritant
Disodium metasilicate pentahydrate, TECHNICAL
Catalog # 28092.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Disodium metasilicate pentahydrate, TECHNICAL
Catalog # 28092.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :212.14 g/mol
Merck Index:13,08716
Melting Pt:72.2 °C
Density:1.749 g/cm³ (20 °C)
EINECS:229-912-9
UN:3253
ADR:8,III
 :GHS05
 :GHS07
CAS Number:10213-79-3
Formula:Na₂SiO₃·5H₂O
MDL Number:MFCD00149176