Order Entry
Czech Republic
ContactUsLinkComponent
 
Sodium hypophosphite monohydrate ≥98%, GPR RECTAPUR®
Catalog # 27898.297
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium hypophosphite monohydrate ≥98%, GPR RECTAPUR®
Catalog # 27898.297
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 0.1 %
  • Ca (Calcium):
    Max. 0.05 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :105.99 g/mol
Melting Pt:90 °C
Density:0.8 g/cm³ (20 °C)
EINECS:231-669-9
CAS Number:10039-56-2
Formula:NaH₂PO₂·H₂O
MDL Number:MFCD06252379