Order Entry
Czech Republic
ContactUsLinkComponent
Hexachloroplatinic(IV) acid hexahydrate ≥37.50% (Pt basis) ACS, Fluka™
 
 
 
Hexachloroplatinic(IV) acid hexahydrate ≥37.50% (Pt basis) ACS, Fluka™
  206083-1G
 :  
 
 
 
Hexachloroplatinic(IV) acid hexahydrate ≥37.50% (Pt basis) ACS, Fluka™
  206083-1G
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    complying
  • Platinum (Pt):
    Min. 37.50 %
  • Solubility in C₂H₅OH:
    complying
  • Alkalis (as Sulfates):
    Max. 0.05 %
  • Application test:
    complying

 

 

 :
 :517.9 g/mol
 :13,07609
 :150 °C
 :2,434 g/cm³ (20 °C)
 :Ambient
 :241-010-7
 :2507
 :8,III
 :GHS05
 :GHS08
 :GHS06
 :18497-13-7
 :H₂PtCl₆·6H₂O
 :MFCD00149910