Order Entry
Czech Republic
ContactUsLinkComponent
Hexachloroplatinic(IV) acid hexahydrate for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Health
Hexachloroplatinic(IV) acid hexahydrate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Corrosive
Health
Hexachloroplatinic(IV) acid hexahydrate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Melting Point:
    150 °C
  • Molar Mass:
    517.94 g/mol
  • Density:
    4 g/cm3 (20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together