

Dodecamolybdophosphoric acid hydrate, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:51429-74-4
:
Formula:H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number:MFCD00149913
:1843.27 g/mol
Merck Index:12,07498
Melting Pt:78…90 °C
Density:1.62 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:234-713-5
UN:3085
ADR:5.1 (8),III
:GHS03
:GHS05
Specification Test Results
- Heavy metals (as Pb):Max. 50 ppm
- Insolubility in Water:Max. 100 ppm
- Cl (Chloride):Max. 50 ppm
- NH4 (Ammonium):Max. 30 ppm
- SO4 (Sulphate):Max. 100 ppm
- Ca (Calcium):Max. 100 ppm
- Cu (Copper):Max. 10 ppm
- Fe (Iron):Max. 20 ppm
- Pb (Lead):Max. 20 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph. Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Informace o balení
Frequently Bought Together