(R)-(-)-3-Chloro-1,2-propanediol, Sigma-Aldrich®
Catalog # SIAL540056-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:110.54 g/mol
Boiling Pt:74…76 °C (15 mmHg)
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Refrigerator
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:435-560-9
UN:2689
CAS Number:57090-45-6
Formula:ClCH₂CHOHCH₂OH
MDL Number:MFCD00135169