

Sodium diethyldithiocarbamate trihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:20624-25-3
:
Formula:(C₂H₅)₂NCSSNa·3H₂O
MDL Number:MFCD00150617
:225.31 g/mol
Merck Index:12,03443
Melting Pt:95…98.5 °C
Density:1.1 g/cm³ (20 °C)
EINECS:205-710-6
ADR:9,III
:M-Clarity™ quality level = MQ200
UN:3077
:GHS07
:GHS09
Frequently Bought Together