Order Entry
Czech Republic
ContactUsLinkComponent
Sodium diethyldithiocarbamate trihydrate ≥97% analytical reagent
Irritant
Environment
Sodium diethyldithiocarbamate trihydrate ≥97% analytical reagent
Catalog # 83864.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Environment
Sodium diethyldithiocarbamate trihydrate ≥97% analytical reagent
Catalog # 83864.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • Ignition residue (SO4):
    30.0 - 35.0 %
  • Suited for copper reagent:
    Passes test

Specification Test Results

About this item

 :
 :225.31 g/mol
Merck Index:12,03443
Melting Pt:95…98,5 °C
Density:1.1 g/cm³ (20 °C)
EINECS:205-710-6
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:20624-25-3
Formula:(C₂H₅)₂NCSSNa·3H₂O
MDL Number:MFCD00150617