Order Entry
Czech Republic
ContactUsLinkComponent
Silver nitrate 1 mol/l (1 N) in aqueous solution volumetric solution
Silver nitrate 1 mol/l (1 N) in aqueous solution volumetric solution
Katalogové číslo REGEN21000W
Supplier:  REAGECON
CAS Number:  
undefined
Silver nitrate 1 mol/l (1 N) in aqueous solution volumetric solution
Katalogové číslo REGEN21000W
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3264
ADR:8,II
 :GHS05
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414