Order Entry
Czech Republic
ContactUsLinkComponent
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol
Catalog # VWRS2715.9011
Supplier:  VWR Collection
CAS Number:  
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol
Catalog # VWRS2715.9011
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:3286
ADR:3,II
 :GHS02
 :GHS06
 :GHS05
 :GHS08
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553