Order Entry
Czech Republic
ContactUsLinkComponent
Sodium chloride 5 mol/l (5 N) in aqueous solution, Sigma-Aldrich®
Sodium chloride 5 mol/l (5 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIALS5150-1L
Supplier:  Merck
CAS Number:  
Sodium chloride 5 mol/l (5 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIALS5150-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :58.44 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477