Order Entry
Czech Republic
ContactUsLinkComponent
Pesticide reference standard, 2,4-Dinitrophenol 5,000 µg/ml in methanol, Supelco®
Pesticide reference standard, 2,4-Dinitrophenol 5,000 µg/ml in methanol, Supelco®
Catalog # SUPL40057
Supplier:  Merck
CAS Number:  
Pesticide reference standard, 2,4-Dinitrophenol 5,000 µg/ml in methanol, Supelco®
Catalog # SUPL40057
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    51-28-5
  • Description:
    2,4-Dinitrophenol 5,000 µg/ml in methanol
  • Formula:
    C₆H₄N₂O₅
  • Description:
    2,4-Dinitrophenol

Specifications

About this item