VWR® ARISTAR®, Zinc Standard Solution, 10000 mg/L Zn in dil. Nitric Acid (from Zn), Standard for ICP
Supplier: VWR Chemicals
Certificates
About this item
Zn in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.
Related Information
Certificates
Search by Lot Number125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125) 500 ml Plastic bottle for liquids DIN 28 (ZPACK15150500)
Informace o balení
VWR® for Calibrations
Produktová literatura
Frequently Bought Together