Zinc standard solution, 1000 mg/L Zn in 0,5 M nitric acid (from Zn(NO₃)₂) Certipur® standard for AAS, Supelco®
Catalog # 1.19806.0100
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Zinc
- Symbol:Zn
Specifications
About this item
Certifications: Traceable to SRM from NIST.