Order Entry
Czech Republic
ContactUsLinkComponent
 
VWR® ARISTAR®, Rubidium Standard Solution, 1000 mg/L Rb in dil. Nitric Acid (from RbNO3), Standard for ICP
  455902G
 :  
undefined
VWR® ARISTAR®, Rubidium Standard Solution, 1000 mg/L Rb in dil. Nitric Acid (from RbNO3), Standard for ICP
  455902G
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Rubidium
  • Symbol:
    Rb

 

 

RbNO₃ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with Certificate of Analysis.