About this item
Ni in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Related Information
Certificates
Search by Lot Number125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125) 500 ml Plastic bottle for liquids DIN 28 (ZPACK15150500)
Packaging Information
SDS_EN_CH_VWRC45572.pdf SDS_EN_DE_VWRC45572.pdf SDS_EN_NL_VWRC45572.pdf SDS_EN_UK_VWRC45572.pdf sds_VWRC45572
SDS
VWR® for Calibrations VWR® Trace Analysis.pdf