Order Entry
Czech Republic
ContactUsLinkComponent
VWR® ARISTAR®, Gallium Standard Solution, 10000 mg/L Ga in dilute Nitric Acid (from Ga), Standard for ICP
VWR® ARISTAR®, Gallium Standard Solution, 10000 mg/L Ga in dilute Nitric Acid (from Ga), Standard for ICP
Catalog # 455392G
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Gallium Standard Solution, 10000 mg/L Ga in dilute Nitric Acid (from Ga), Standard for ICP
Catalog # 455392G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Gallium
  • Symbol:
    Ga

Specifications

About this item

Ga in HNO₃.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.