Order Entry
Czech Republic
ContactUsLinkComponent
Beryllium standard solution, 1000 mg/L Be in 0,5 M nitric acid (from BeO.(Be(OAc)₂)₃) Certipur® standard for AAS, Supelco®
Beryllium standard solution, 1000 mg/L Be in 0,5 M nitric acid (from BeO.(Be(OAc)₂)₃) Certipur® standard for AAS, Supelco®
Catalog # 1.70207.0100
Supplier:  Merck
CAS Number:  
Beryllium standard solution, 1000 mg/L Be in 0,5 M nitric acid (from BeO.(Be(OAc)₂)₃) Certipur® standard for AAS, Supelco®
Catalog # 1.70207.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Beryllium
  • Symbol:
    Be

Specifications

About this item

Certifications: Traceable to SRM from NIST.