Order Entry
Czech Republic
ContactUsLinkComponent
DL-Lysine monohydrate 97%
DL-Lysine monohydrate 97%
Catalog # ACRO365650250
CAS Number:  
undefined
DL-Lysine monohydrate 97%
Catalog # ACRO365650250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    7 % to 12 % (105°C, 2 hrs)
  • Titration with HClO4:
    ≥96.0 %

Specification Test Results

About this item

 :
 :164.2 g/mol
EINECS:200-740-6
CAS Number:885701-25-7
Formula:C₆H₁₄N₂O₂·1H₂O
MDL Number:MFCD00151036