Order Entry
Czech Republic
ContactUsLinkComponent
 
 
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%
 
undefined
 
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4320 to 1.4360 (20°C, 589 nm)
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together