

Titanium(III) chloride 30% in hydrochloric acid 10% for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7705-07-9
:
Formula:Cl₃Ti
MDL Number:MFCD00011266
:154.24 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:231-728-9
UN:1789
:GHS05
:GHS07
Specification Test Results
- Density:1.017 g/cm3 (20 °C)
- pH:0.5 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Frequently Bought Together