Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butylchlorodimethylsilane 50% (w/w) in toluene
Flammable
Health
Corrosive
Irritant
tert-Butylchlorodimethylsilane 50% (w/w) in toluene
Catalog # B21286.18
CAS Number:  
Flammable
Health
Corrosive
Irritant
tert-Butylchlorodimethylsilane 50% (w/w) in toluene
Catalog # B21286.18
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

CAS Number:18162-48-6
 :
Formula:C₆H₁₅ClSi
MDL Number:MFCD00000501
 :150.72 g/mol
Density:0.87
Flash Pt:7 °C (45 °F)
Storage Temperature:Ambient
EINECS:242-042-4
UN:2924
ADR:3,II
 :GHS02
 :GHS08
 :GHS05
 :GHS07