Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Rifampicin

Rifampicin

Catalog #
Rifampicin
Catalog #
Supplier:  
CAS Number:  13292-46-1
Rifampicin
CAS Number:  13292-46-1
Product Details & Documents
Synonyms:  Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
  • CAS Number:13292-46-1
  • Formula:C₄₃H₅₈N₄O₁₂
  • MDL Number:MFCD00151389
  • Molecular Weight:822.95 g/mol
  • Merck Index:12,08382
  • Melting Pt:183…188 °C
  • EINECS:236-312-0