Glyoxal 40% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-22-2
:
Formula:C₂H₂O₂
MDL Number:MFCD00006957
:58.04 g/mol
Density:1.27 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:203-474-9
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together