Order Entry
Czech Republic
ContactUsLinkComponent
Petroleum spirit, 30…60 °C, anhydrous, Sigma-Aldrich®
Flammable
Health
Irritant
Environment
Petroleum spirit, 30…60 °C, anhydrous, Sigma-Aldrich®
Catalog # SIAL300314-100ML
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
Environment
Petroleum spirit, 30…60 °C, anhydrous, Sigma-Aldrich®
Catalog # SIAL300314-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, Sure/Seal
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Boiling Pt:35…60 °C (1013 hPa)
Storage Temperature:Ambient
EINECS:265-151-9
ADR:3,II
 :M-Clarity™ quality level = MQ100
UN:1268
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849