Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyanine 647-dUTP
Cyanine 647-dUTP, lyophilised blue solid
Cyanine 647-dUTP, lyophilised blue solid
Cyanine 647-dUTP, lyophilised blue solid
Catalog # 40065.
Supplier:
Biotium
CAS Number:
Expand image
Cyanine 647-dUTP, lyophilised blue solid
Catalog # 40065.
Supplier:
Biotium
CAS Number:
Specifications
Pack type:
Plastic vial
Pk:
25 Nmo
Seq:
0311
Specifications
About this item
Product Details
:
:
~1151 g/mol
Storage Temperature:
Freezer
Product Details