Order Entry
Czech Republic
ContactUsLinkComponent
Zimmermann-Reinhardt's solution
Corrosive
Zimmermann-Reinhardt's solution
Catalog # 9110.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Zimmermann-Reinhardt's solution
Catalog # 9110.1000
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :223.06 g/mol
UN:3264
ADR:8,II
 :GHS05