VWR® AVS TITRINORM, Zinc Standard Solution, 1000 mg/L Zn in dil. Nitric Acid, Standard for AAS
Supplier: VWR Chemicals
Certificates
About this item
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%
Related Information
Certificates
Search by Lot NumberSDS_EN_CH_VWRC86725.pdf SDS_EN_DE_VWRC86725.pdf SDS_EN_NL_VWRC86725.pdf SDS_EN_UK_VWRC86725.pdf sds_VWRC86725
SDS
VWR® for Calibrations
Produktová literatura
Frequently Bought Together