Patent blue V calcium salt, TECHNICAL
Catalog # 34233.128
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:1159.45 g/mol
Melting Pt:200 °C
Storage Temperature:Ambient
EINECS:222-573-8
CAS Number:3536-49-0
Formula:C₅₄H₆₂CaN₄O₁₄S₄
MDL Number:MFCD00038746