Order Entry
Czech Republic
ContactUsLinkComponent
Congo red, TECHNICAL
Health
Congo red, TECHNICAL
Catalog # 34140.184
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Congo red, TECHNICAL
Catalog # 34140.184
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :696.67 g/mol
Merck Index:12,02562
Melting Pt:> 360 °C
EINECS:209-358-4
 :GHS08
Storage Temperature:Ambient
CAS Number:573-58-0
Formula:C₃₂H₂₂N₆Na₂O₆S₂
MDL Number:MFCD00004028